Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Sigma-Aldrich Acid Yellow 17, Powder, 60% Dye Content, 100 g

Catalog No. NC3846305 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred

Acid Yellow 17 (C.I. 18965) is a synthetic, anionic azo dye characterized by a stable aromatic structure containing sulfonic acid and hydroxyl functional groups. This powder reagent is utilized across clinical and diagnostic applications, including hematology and histology, as well as in industrial sectors such as textiles and detergents.

  • Dye content is 60%.
  • Molecular weight of 551.29 g/mol.
  • Empirical formula of C16H10Cl2N4Na2O7S2.
  • Maximum absorbance (λmax) at 400 nm.
  • Extinction coefficient of 8000 at 252-258 nm in water at 0.02 g/L.
  • Storage at room temperature.
  • Classified as a combustible solid (storage class 11).

Catalog No. NC3846305 Supplier Sigma Organic Chemistry Supplier No. 201987100G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Acid Yellow 17 (C.I. 18965) is a synthetic, anionic azo dye characterized by a stable aromatic structure containing sulfonic acid and hydroxyl functional groups. This powder reagent is utilized across clinical and diagnostic applications, including hematology and histology, as well as in industrial sectors such as textiles and detergents.

  • Dye content is 60%.
  • Molecular weight of 551.29 g/mol.
  • Empirical formula of C16H10Cl2N4Na2O7S2.
  • Maximum absorbance (λmax) at 400 nm.
  • Extinction coefficient of 8000 at 252-258 nm in water at 0.02 g/L.
  • Storage at room temperature.
  • Classified as a combustible solid (storage class 11).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.