Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Fine Chemicals Biosciences Sigma-Aldrich BioReagent, 5(6)-Carboxy-2′,7′-dichlorofluorescein, ≥95% (TLC), Suitable for Fluorescence

Catalog No. NC3984058
Encompass

5(6)-Carboxy-2′,7′-dichlorofluorescein may be used as a specific substrate/probe to study the identity, activity and specificity of multidrug resistance protein 2 (MRP2) molecules. It is also suitable as a pH-indicator. It functions as a fluorescent intracellular pH indicator in the pH 4–5 range.

  • Assay: ≥95% (TLC)
  • Form: Solid
  • Solubility: DMF: soluble, DMSO: soluble, H2O: soluble (pH ≥ 5)
  • Fluorescence: λex 504 nm; λem 529 nm in 0.1 M Tris pH 8.0
  • Suitability: Suitable for fluorescence
  • Empirical formula (Hill Notation): C21H10Cl2O7
  • CAS number: 111843-78-8
  • Molecular weight: 445.21

Catalog No. NC3984058 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 21882100MGF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5(6)-Carboxy-2′,7′-dichlorofluorescein may be used as a specific substrate/probe to study the identity, activity and specificity of multidrug resistance protein 2 (MRP2) molecules. It is also suitable as a pH-indicator. It functions as a fluorescent intracellular pH indicator in the pH 4–5 range.

  • Assay: ≥95% (TLC)
  • Form: Solid
  • Solubility: DMF: soluble, DMSO: soluble, H2O: soluble (pH ≥ 5)
  • Fluorescence: λex 504 nm; λem 529 nm in 0.1 M Tris pH 8.0
  • Suitability: Suitable for fluorescence
  • Empirical formula (Hill Notation): C21H10Cl2O7
  • CAS number: 111843-78-8
  • Molecular weight: 445.21

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.