Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Sigma-Aldrich Bromoethane For Synthesis, Liquid, ≥99.0% GC, 250 mL

Catalog No. NC3865988 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred

Bromoethane, also known as ethyl bromide, is a liquid reagent supplied for chemical synthesis applications. This compound features a molar mass of 108.97 g/mol and is provided with a high assay of at least 99.0% by GC analysis.

  • Formula: C<sub>2</sub>H<sub>5</sub>Br
  • Assay: ≥ 99.0% (GC)
  • Boiling point: 37–39 °C
  • Melting point: -119 °C
  • Density: 1.460 g/cm<sup>3</sup>
  • Flash point: -23 °C
  • Storage temperature: 2–30 °C
  • Hazard profile: Highly flammable liquid and vapour, harmful if swallowed or inhaled, and suspected of causing cancer

Catalog No. NC3865988 Supplier Sigma Organic Chemistry Supplier No. 8008710250
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bromoethane, also known as ethyl bromide, is a liquid reagent supplied for chemical synthesis applications. This compound features a molar mass of 108.97 g/mol and is provided with a high assay of at least 99.0% by GC analysis.

  • Formula: C<sub>2</sub>H<sub>5</sub>Br
  • Assay: ≥ 99.0% (GC)
  • Boiling point: 37–39 °C
  • Melting point: -119 °C
  • Density: 1.460 g/cm<sup>3</sup>
  • Flash point: -23 °C
  • Storage temperature: 2–30 °C
  • Hazard profile: Highly flammable liquid and vapour, harmful if swallowed or inhaled, and suspected of causing cancer

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.