Learn More
Sigma Aldrich Fine Chemicals Biosciences Sigma-Aldrich Dialysis Kit, 10-250 μL, 12-14 kDa MWCO, 50 Tubes
Catalog No. NC3985630
Encompass
The Pur-A-Lyzer Dialysis Kits feature innovative tubing for convenient pipette tip loading and unloading, ensuring 98% sample recovery. Designed for high-throughput, they accommodate many samples in a single buffer tank, enabling rapid, contamination-free extraction of macromolecules for protein purification, sequencing, peptide mapping, and amino acid analysis.
  • Achieves 98% sample recovery
  • Enables high-throughput dialysis
  • Suitable for purifying proteins, sequencing, peptide mapping
  • Effective for dialysis or buffer exchange (10 mL to 20 mL)
  • Removes salts, surfactants, and contaminating micromolecules
Catalog No. NC3985630 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. PURN120501KT
The Pur-A-Lyzer Dialysis Kits feature innovative tubing for convenient pipette tip loading and unloading, ensuring 98% sample recovery. Designed for high-throughput, they accommodate many samples in a single buffer tank, enabling rapid, contamination-free extraction of macromolecules for protein purification, sequencing, peptide mapping, and amino acid analysis.
  • Achieves 98% sample recovery
  • Enables high-throughput dialysis
  • Suitable for purifying proteins, sequencing, peptide mapping
  • Effective for dialysis or buffer exchange (10 mL to 20 mL)
  • Removes salts, surfactants, and contaminating micromolecules