Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Sigma-Aldrich™ Flash Silica (Dried, Spherical)

Catalog No. 111008788 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Includes:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
11-100-8788 1 kg
11-101-0960 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-8788 Supplier MilliporeSigma™ Supelco™ Supplier No. 97729U
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

High-purity grade

  • Pore Size: 0.7-0.9 cm3/g pore volume
  • Surface Area: 480 m2/g
  • MDL Number: MFCD00011232
  • BP: 2230°C
  • MP: gt;1600°C
  • CAS: 112926-00-8
TRUSTED_SUSTAINABILITY

Specifications

Includes 1 kg
For Use With (Application) Flash Chromatography
Product Line Sigma-Aldrich

Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.