Learn More
Description
- When blood films are stained using Giemsa Stain, the nucleus and cytoplasm of white blood cells take on characteristic blue or pink coloration
- The use of purified eosin and thiazine dyes minimizes lot-to-lot variation
Specifications
Specifications
| Quantity | 500mL |
| Storage Requirements | 18-26°C |
| Format | Solution |
| Concentration | 0.4% (w/v) in buffered methanol solution, pH 6.8, with stabilizers |
| For Use With (Application) | For use in staining blood films or bone marrow films |
| Color | Faint Blue to Dark Blue |
| pH | 6.8 |
| Synonym | Giemsa Solution |
| Product Type | Stain |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.