Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Reticulocyte Stain

Catalog No. SDR4132
Encompass
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SDR4132 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SDR4132 Supplier Sigma Diagnostics™ Supplier No. R4132120ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for the identification of reticulocytes in blood films

  • Blood and reticulocyte staining solution (New Methylene Blue) are mixed and incubated briefly at room temperature
  • Wedge or spun smears are made on microscope slides, air dried and evaluated under oil immersion on a light microscope
  • A red blood cell scoring positive would be observed containing two or more blue-stained granules
TRUSTED_SUSTAINABILITY

Specifications

Quantity 120 mL
Storage Requirements 18–26°C
Format Solution
For Use With (Application) For the identification of reticulocytes in blood films.
Product Type Reticulocyte Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.