Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Eosin Y Solution, alcoholic

Catalog No. SDHT110132
Encompass
Change view
Click to view available options
Quantity:
500mL
1L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SDHT110132 1L
SDHT110116 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SDHT110132 Supplier Sigma Diagnostics™ Supplier No. HT1101321L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

General-purpose cytoplasmic counterstain

  • Certified Eosin Y, 0.5% (w/v) in acidified 90% ethanol
  • For in vitro diagnostic use
TRUSTED_SUSTAINABILITY

Specifications

Color Bright Pink to Pinkish-Orange
Format Solution
For Use With (Application) Used with hematoxylin and eosin staining
Quantity 1L
Product Type Eosin Y solution, alcoholic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.