Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Eosin Y Solution, aqueous

Catalog No. SDHT110216
Encompass
Change view
Click to view available options
Quantity:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SDHT110216 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SDHT110216 Supplier Sigma Diagnostics™ Supplier No. HT110216500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

General purpose cytoplasmic counterstain

  • Certified Eosin Y, 0.5% (w/v) in water. Not acidified.
TRUSTED_SUSTAINABILITY

Specifications

Color Greenish-Pink-Orange
Format Solution
For Use With (Application) Used with hematoxylin and eosin staining
Quantity 500mL
Storage Requirements 18–26°C
Product Type Aqueous Eosin Y Stain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.