Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Gold chloride solution

Catalog No. SDHT1004
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SDHT1004 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SDHT1004 Supplier Sigma Diagnostics™ Supplier No. HT1004100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used for the toning of sections stained with the Silver Methanamine procedure

  • For use in Silver Stain (Modified GMS) Sigma Procedure No. HT100
  • 200 mg/dL in deionized water

For in vitro diagnostic use

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Format Solution
For Use With (Application) For use in silver stain (modified GMS) Sigma procedure No. HT100
Quantity 100 mL
Storage Requirements 2° to 8°C
Product Type Gold chloride solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.