Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Reticulum Stain Kit

Catalog No. SDHT102A
Encompass
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SDHT102A 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SDHT102A Supplier Sigma Diagnostics™ Supplier No. HT102A1KT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Ferric Ammonium Sulfate Solution (kit only), 500mL
  • Gold Chloride Solution (kit only), 100mL
  • Oxalic Acid Solution (kit only), 100mL
  • Potassium Permanganate Solution (kit only), 100mL
  • Silver Nitrate Solution (kit only), 50mL
  • Sodium Hydroxide Solution (kit only), 100mL
  • Sodium Thiosulfate Solution (kit only), 500mL

Intended to demonstrate reticular fibers in tissue sections

  • Based on the method of Gordon and Sweets
  • For in vitro diagnostic use
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Product Type Staining Reagent
Quantity 1 kit
For Use With (Application) Intended to demonstrate reticular Fibers in paraffin sections
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.