Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Histology Stains and Reagents - Schiff's Reagent

Catalog No. 01288072 Shop All MilliporeSigma Supelco Products
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-288-072 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-288-072 Supplier Sigma Diagnostics™ Supplier No. 3952016500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the demonstration of lymphocytes and mucopolysaccharides

  • For use in Periodic Acid Schiff (PAS) Sigma Procedure No. 395

Applications:

For in vitro diagnostic use

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Format Solution
For Use With (Application) For use with blood, bone marrow, tissue touch preparations or routine tissue sections.
Quantity 500 mL
Storage Requirements 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.