Learn More
Sigma Diagnostics™ WeigertÆs iron hematoxylin solution
Catalog No. SDHT1079
Encompass
Change view
Click to view available options
Quantity:
1 Set
Catalog No. Quantity
SDHT1079 1 Set
1 options
Catalog No. SDHT1079 Supplier Sigma Diagnostics™ Supplier No. HT10791SET
Includes
  • Weigert's iron hematoxylin solution, Part A 500mL
  • Weigert's iron hematoxylin solution, Part B 500mL
Resists decolorization better than aluminum based hematoxylin formulations
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Lengthy or acidic stain procedures
Includes
  • Weigert’s iron hematoxylin solution, Part A 500mL
  • Weigert’s iron hematoxylin solution, Part B 500mL
Quantity 1 Set