Learn More
Sigma Diagnostics™ Papanicolaou Stains
Catalog No. SDHT40132
Encompass
Change view
Click to view available options
OG-6; 1L; Orange
Catalog No.
SDHT40132 OG-6; 1L; Orange
1 options
This item is not returnable. View return policy
Catalog No. SDHT40132 Supplier Sigma Diagnostics™ Supplier No. HT401321L
This item is not returnable. View return policy
For routine diagnostic cytology to aid in identification and classification of exfoliative cells
  • Orange G (certified), 0.3% and phosphotungstic acid, 0.015%, in denatured alcohol