Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Sigma Aldrich Poly(ethylene Glycol), Average Mn 950-1,050, Waxy Solid, 250 g

Catalog No. NC4000705 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred

Poly(ethylene glycol) is a non-toxic and water-soluble polymer characterized by its ability to resist immune system recognition. This biodegradable compound serves as a versatile ingredient in pharmaceutical and cosmetic formulations, offering a sustainable profile derived from renewable resources.

  • Average Mn 950-1,050
  • Form: waxy solid
  • Solubility: 800 mg/mL in water at 20 °C
  • Density: 1.0926 g/mL
  • Viscosity: 17.4 cSt
  • Melting point: 39 °C
  • Chemical formula: H(OCH2CH2)nOH
  • CAS number: 25322-68-3

Catalog No. NC4000705 Supplier Sigma Organic Chemistry Supplier No. P3515250G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Poly(ethylene glycol) is a non-toxic and water-soluble polymer characterized by its ability to resist immune system recognition. This biodegradable compound serves as a versatile ingredient in pharmaceutical and cosmetic formulations, offering a sustainable profile derived from renewable resources.

  • Average Mn 950-1,050
  • Form: waxy solid
  • Solubility: 800 mg/mL in water at 20 °C
  • Density: 1.0926 g/mL
  • Viscosity: 17.4 cSt
  • Melting point: 39 °C
  • Chemical formula: H(OCH2CH2)nOH
  • CAS number: 25322-68-3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.