Learn More
Sigma Diagnostics™ Poly-Prep Slides
Catalog No. SDP0425
Encompass
Change view
Click to view available options
Quantity:
72/Pk.
Catalog No. Quantity
SDP0425 72/Pk.
1 options
Catalog No. SDP0425 Supplier Sigma Diagnostics™ Supplier No. P042572EA
Coated with poly-L-lysine for tissue section adhesion in routine histological uses
  • Smooth edges, free from chips and cracks
  • Clear glass with white frosted ends
  • Comes in a pack of 72
TRUSTED_SUSTAINABILITY
Specifications
Frosted Yes
Material Glass
Coating Poly-L-Lysine
Quantity 72/Pk.