Learn More
Sigma Diagnostics™ Silane-Prep Slides
Catalog No. SDS4651
Encompass
Change view
Click to view available options
Quantity:
72/Pk.
Catalog No. Quantity
SDS4651 72/Pk.
1 options
Catalog No. SDS4651 Supplier Sigma Diagnostics™ Supplier No. S465172EA
Coated with silane (aminoalkylsilane)
  • Clear glass slide with frosted end for slide/patient identification
  • Smooth edges free of chips or cracks
  • Tested for tissue adherence performance
  • Comes in a pack of 72
TRUSTED_SUSTAINABILITY
Specifications
Frosted 1 End
Coating Silane
Quantity 72/Pk.