Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Sigma-Aldrich™ Silica Gel, Technical Grade, 200-425 mesh

Catalog No. 111012523 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2523 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2523 Supplier MilliporeSigma™ Supelco™ Supplier No. 6455245KG

missing translation for 'validMainframeMsg'

Residual water (≤12%). For preparative liquid chromatography.

Silica gel is exclusively used as a support material for the active titanium (III) centers in Ziegler-Natta catalysts. Silica gel allows a controlled fragmentation leading to form uniform polymer particles with narrow particle size distribution and high bulk density.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 200/425 Mesh
Pore Size 60 Å
Grade Technical Grade
Packaging Fiber Drum
pH 6.5 to 7.5
Quantity 5 kg
Surface Area (Metric) 500 to 600 m2/g (approx.)
Product Line Sigma-Aldrich
Physical Form Powder
MDL Number MFCD00011232
CAS 112926-00-8
Show More Show Less

Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.