Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Sigma-Aldrich™ Silica Gel, Technical Grade (with Ca, ∼0.1), 230-400 mesh

Catalog No. 111012541 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2541 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2541 Supplier MilliporeSigma™ Supelco™ Supplier No. 124795KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For preparative liquid chromatography

Silica gel, highly porous textured matrix are generally used for synthesis of adsorbents. They are corpuscular in structure. Silica surface possess a layer of hydroxyl groups and adsorbed water molecules, which can be removed by drying at high temperature or even by treatment with suitable reagents. Silica gel surface plays important role in catalysis, chemical separation and microelectronic fabrications.
TRUSTED_SUSTAINABILITY

Specifications

Particle Size 230/400 Mesh
Pore Size 60 Å
Grade Technical Grade (w/ Ca, ∼0.1%)
Quantity 5 kg
Surface Area (Metric) 500 m2/g (approx.)
Product Line Sigma-Aldrich
Description Ca, 0.1 to 0.3%
Physical Form Powder
MDL Number MFCD00011232
CAS 112926-00-8

Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.