Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Sigma-Aldrich SPGS-550-M, 2% w/w in H2O, Liquid, 50 mL

Catalog No. NC3877669 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass

SPGS-550-M is a phytosterol-based amphiphile designed as a third-generation designer surfactant for use in transition-metal-catalyzed cross-coupling reactions in water at room temperature. Provided as a 2% w/w liquid in water, this product offers an energy-efficient, greener alternative for chemical synthesis in research and laboratory settings.

  • Empirical formula C58H104O16
  • Molecular weight of 1057.44 g/mol
  • Concentration of 2% w/w in H2O
  • Density of 1.005 g/mL
  • Refractive index of n20/D 1.336
  • Storage temperature of 2-8°C
  • Suitable for room temperature cross-coupling reactions

Catalog No. NC3877669 Supplier Sigma Organic Chemistry Supplier No. 77603350ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SPGS-550-M is a phytosterol-based amphiphile designed as a third-generation designer surfactant for use in transition-metal-catalyzed cross-coupling reactions in water at room temperature. Provided as a 2% w/w liquid in water, this product offers an energy-efficient, greener alternative for chemical synthesis in research and laboratory settings.

  • Empirical formula C58H104O16
  • Molecular weight of 1057.44 g/mol
  • Concentration of 2% w/w in H2O
  • Density of 1.005 g/mL
  • Refractive index of n20/D 1.336
  • Storage temperature of 2-8°C
  • Suitable for room temperature cross-coupling reactions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.