Learn More
Sigma Aldrich Fine Chemicals Biosciences Sigma AldrichR micro pH co1EA
Catalog No. 502904090
Encompass
Designed with a Refillable Ag/AgCl reference half cell. These electrodes feature ultra thin insertion stems and miniature pH bulbs for measurement of the smallest of samples. Depends on the length the extra long insertion stem allows this combination electrode to measure samples upto the bottom of 7 in. long NMR tubes.Sturdy and versatile probe for measuring small samples in cuvets and small lab glassware 0-14 pH.Attention These electrodes come with a little white plastic transport cap attached to the end please remove before use.Temperature Range-5 to 100C (for continuous use)-5 to 110C (for intermittent use)
Catalog No. 50-290-4090 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E52591EA
Designed with a Refillable Ag/AgCl reference half cell. These electrodes feature ultra thin insertion stems and miniature pH bulbs for measurement of the smallest of samples. Depends on the length the extra long insertion stem allows this combination electrode to measure samples upto the bottom of 7 in. long NMR tubes.Sturdy and versatile probe for measuring small samples in cuvets and small lab glassware 0-14 pH.Attention These electrodes come with a little white plastic transport cap attached to the end please remove before use.Temperature Range-5 to 100C (for continuous use)-5 to 110C (for intermittent use)