Learn More
Sigma Aldrich Fine Chemicals Biosciences Sigma AldrichR vacuum filt1EA
Catalog No. 502905786
Encompass
Vacuum filtration apparatus for HPLC solvent purification particulate analysis bacteriological studies radiochemical determinations quantitative gravimetric analysis and similar applications. Clamp seals filter membrane between flanges on funnel top and funnel stem.Includes funnel top funnel stem/membrane support aluminum clamp stopper (except standard taper units) and filtration flask (where indicated).for replacement parts visit our online glassware catalog at www.sigma-aldrich.com
Catalog No. 50-290-5786 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z2905051EA
Vacuum filtration apparatus for HPLC solvent purification particulate analysis bacteriological studies radiochemical determinations quantitative gravimetric analysis and similar applications. Clamp seals filter membrane between flanges on funnel top and funnel stem.Includes funnel top funnel stem/membrane support aluminum clamp stopper (except standard taper units) and filtration flask (where indicated).for replacement parts visit our online glassware catalog at www.sigma-aldrich.com