Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Bouin's Solution

Catalog No. SDHT10132
Encompass
Change view
Click to view available options
Quantity:
1L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SDHT10132 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SDHT10132 Supplier Sigma Diagnostics™ Supplier No. HT101321L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fixative for preserving soft and delicate structures

  • Prepared with saturated picric acid, formaldehyde and acetic acid
  • Composition: acetic acid, 5%; formaldehyde, 9%; picric acid, 0.9%
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Format Solution
For Use With (Application) Used as a mordant in various trichrome procedures
Quantity 1L
Synonym Bouin’s fluid
Product Type Bouin’s solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.