Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Histopaque™-1077 Cell Separation Medium

Catalog No. SD10771A
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SD10771A 100 mL
SD10771B 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SD10771A Supplier Sigma Diagnostics™ Supplier No. 10771100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Facilitates recovery of large numbers of viable lymphocytes and other mononuclear cells from small volumes of whole blood. Sigma-Aldrich™ Histopaque™-1077 is suitable for human lymphocyte antigen (HLA) typing and as the initial isolation step prior to enumeration of T, B, and 'null' lymphocytes.

  • Clear, colorless liquid
  • Contains polysucrose and sodium diatrizoate
  • Sterile-filtered
  • Endotoxin tested
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
For Use With (Application) For ease of cell recovery
Endotoxin Level ≤0.3EU/mL
pH 8.8 to 9.0
Product Type Centrifugation Media
Density 1.077 g/mL
Sterility Sterile
Form Liquid
Content And Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.