Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Diagnostics™ Naphthol AS-D Chloroacetate (Specific Esterase) Kit

Catalog No. SD91C
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SD91C 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SD91C Supplier Sigma Diagnostics™ Supplier No. 91C1KT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Citrate Solution, pH 3.6 ± 0.1 (25°C), 27mM 50mL
  • Fast Red Violet LB Base solution 10mL
  • Naphthol AS-D Chloroacetate solution 50mL
  • Naphthol AS-D Chloroacetate solution 50mL

Used to detect neutrophils in peripheral blood, marrow or tissue sections embedded in paraffin

  • With the 91C kit, Naphthol AS-D Chloroacetate is enzymatically hydrolyzed by “specific esterase, ” liberating a free naphthol compound
  • This couples with a diazonium compound, forming highly colored deposits at sites of enzyme activity
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) To detect neutrophils in peripheral blood, marrow or tissue sections embedded in paraffin
Includes
  • Citrate Solution, pH 3.6 ± 0.1 (25°C), 27mM 50mL
  • Fast Red Violet LB Base solution 10mL
  • Naphthol AS-D Chloroacetate solution 50mL
  • Naphthol AS-D Chloroacetate solution 50mL
Quantity 1 Kit
Storage Requirements 2° to 8°C
Synonym Leder Stain
Product Type Naphthol AS-D Chloroacetate (Specific Esterase) Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.