Learn More
Sigma Aldrich Fine Chemicals Biosciences SIGMAFAST BCIP/NBT
Catalog No. NC0995028
Encompass
Synonym: BCIP/NBT Alkaline Phosphatase Substrate. Each tablet dissolved in 10 ml deionized water yields a ready-to-use buffered solution containing BCIP/NBT, pH 9.5.
Catalog No. NC0995028 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B565525TAB
Synonym: BCIP/NBT Alkaline Phosphatase Substrate. Each tablet dissolved in 10 ml deionized water yields a ready-to-use buffered solution containing BCIP/NBT, pH 9.5.