Learn More
Andwin Scientific SIGMA I3146-5ML ITSSIGMA I314
SDP
Catalog No. NC9069158
Encompass
The ITS Liquid Media Supplement is a solution made to be used for cell culture. This product consists of 1.0 mg/ml recombinant human insulin, 0.55 mg/ml human transferrin (substantially iron-free), and 0.5 g/ml sodium selenite at the 100x concentration. This liquid solution is sterile-filtered.
Catalog No. NC9069158 Supplier Andwin Scientific Supplier No. I31465ML
The ITS Liquid Media Supplement is a solution made to be used for cell culture. This product consists of 1.0 mg/ml recombinant human insulin, 0.55 mg/ml human transferrin (substantially iron-free), and 0.5 g/ml sodium selenite at the 100x concentration. This liquid solution is sterile-filtered.