Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sigma-ligand-1 hydrochloride | 139652-86-1 | 99.1% | 472.02 g/mol | C27H34ClNO4 | 5 MG
SDP

Catalog No. 5000245696
Encompass_Preferred

Sigma-LIGAND-1 hydrochloride is the hydrochloride salt provided as an analytical reference standard for use in chromatographic and mass-spectrometry method development, qualitative identification, and quantitative assays (CAS 139652-86-1).

  • Purity of 99.13% suitable for analytical use.
  • Formulated as a reference standard for qualitative and quantitative assays.
  • Compatible with HPLC and LC-MS method development.
  • Molecular weight 472.02 g/mol.
  • Soluble in DMSO for solution preparation.

Catalog No. 50-002-45696 Supplier Medchemexpress LLC Supplier No. HY101626A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sigma-LIGAND-1 hydrochloride is the hydrochloride salt provided as an analytical reference standard for use in chromatographic and mass-spectrometry method development, qualitative identification, and quantitative assays (CAS 139652-86-1).

  • Purity of 99.13% suitable for analytical use.
  • Formulated as a reference standard for qualitative and quantitative assays.
  • Compatible with HPLC and LC-MS method development.
  • Molecular weight 472.02 g/mol.
  • Soluble in DMSO for solution preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.