Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sigma-ligand-1 hydrochloride | 139652-86-1 | MFCD09966150 | 99.1% | 472.02 g·mol⁻1 | C27H34ClNO4 | 25 MG
SDP

Catalog No. 5000248955
Encompass_Preferred

Sigma-LIGAND-1 hydrochloride is the hydrochloride salt of a sigma receptor ligand supplied as a high-purity solid for research use. The product is accompanied by analytical documentation (COA, SDS, H NMR, RP-HPLC, MS) to support quality control and is available in small pack sizes suitable for assay development and medicinal chemistry research.

  • High purity suitable for research applications.
  • Solid, off-white to light yellow physical form.
  • Provided with COA, SDS, H NMR, RP-HPLC, and MS documentation.
  • Available in small pack sizes, including 25 mg.
  • Molecular weight 472.02 g·mol⁻1 and formula C27H34ClNO4.
  • Suitable for in vitro and in vivo assay development.

Catalog No. 50-002-48955 Supplier Medchemexpress LLC Supplier No. HY101626A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sigma-LIGAND-1 hydrochloride is the hydrochloride salt of a sigma receptor ligand supplied as a high-purity solid for research use. The product is accompanied by analytical documentation (COA, SDS, H NMR, RP-HPLC, MS) to support quality control and is available in small pack sizes suitable for assay development and medicinal chemistry research.

  • High purity suitable for research applications.
  • Solid, off-white to light yellow physical form.
  • Provided with COA, SDS, H NMR, RP-HPLC, and MS documentation.
  • Available in small pack sizes, including 25 mg.
  • Molecular weight 472.02 g·mol⁻1 and formula C27H34ClNO4.
  • Suitable for in vitro and in vivo assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.