Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Pseudo™ Corpse Scent (Drowned Victim), MilliporeSigma™ Supelco™

Catalog No. p-7187516 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 Caps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4441 50 Caps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4441 Supplier MilliporeSigma Supelco Supplier No. P718450CAP
Only null left
Add to Cart
Add to Cart

Specifications

Product Line Sigma Pseudo
Physical Form Neat
MDL Number MFCD00165840
Type Standard
Quantity 50 Caps

Sigma Pseudo is a trademark of Sigma-Aldrich Co. LLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.