Learn More
MilliporeSigma™ Supelco™ Sigma TLC spray box
Catalog No. 111016155 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Compatible with Most Sigma Sprays
Catalog No. For Use With (Equipment)
11-101-6155 Compatible with Most Sigma Sprays
1 options
Catalog No. 11-101-6155 Supplier MilliporeSigma™ Supelco™ Supplier No. S15095EA
Add to Cart
Add to Cart
Sigma TLC spray box may be used to spray reagent for TLC plates and may be used in thin layer chromatography and HPTLC.
The reagent/chemical spraying on the TLC plates are generally carried out inside a fume hood or commercially available TLC spray cabinet with blower and exhaust hose. The TLC plate can be placed supported upright or on a paper inside the spray box. Spraying the box ensures complete removal of reagent mist.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) TLC
For Use With (Equipment) Compatible with Most Sigma Sprays
Dimensions (L x W x H) ̃14 in. x 14 in. x 14 in.