Learn More
Sigma Aldrich Fine Chemicals Biosciences SIGMAFAST™ OPD | 5 sets
Catalog No. NC1804657
Encompass
SIGMAFAST™ OPD (o-Phenylenediamine dihydrochloride) tablets have been developed for use as a soluble substrate for the detection of peroxidase activity in enzyme immunoassays (EIA). OPD is the EIA substrate of choice as it exhibits high sensitivity. EIA applications utilizing OPD may be read in timed assays or stopped with dilute acid solutions for delayed readings.
Catalog No. NC1804657 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P91875SET
SIGMAFAST™ OPD (o-Phenylenediamine dihydrochloride) tablets have been developed for use as a soluble substrate for the detection of peroxidase activity in enzyme immunoassays (EIA). OPD is the EIA substrate of choice as it exhibits high sensitivity. EIA applications utilizing OPD may be read in timed assays or stopped with dilute acid solutions for delayed readings.