Learn More
Sigma Aldrich Fine Chemicals Biosciences Sigmatrix Urine Diluent
Catalog No. 502900357
Encompass
Sigmatrix Urine Diluent is a non-biological diluent that mimics human urine.This product can be used as a diluent for sample prep controls and calibrators and is recommended for LC-MS based applications. Sigmatrix Urine Diluent has been successfully used in enzymatic hydrolysis of drug glucuronides and LC-MS analysis of drugs and drug metabolites. The formulation is dye-free.
Catalog No. 50-290-0357 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE007450ML
Sigmatrix Urine Diluent is a non-biological diluent that mimics human urine.This product can be used as a diluent for sample prep controls and calibrators and is recommended for LC-MS based applications. Sigmatrix Urine Diluent has been successfully used in enzymatic hydrolysis of drug glucuronides and LC-MS analysis of drugs and drug metabolites. The formulation is dye-free.