Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (±)-Sigmoidin A | 176046-04-1 | 1 MG
SDP

Catalog No. 5000417348
Encompass_Preferred

The product is an active compound suitable for various biochemical research applications.

  • Available in solid form, off-white to light yellow in color.
  • Purity is 98.46%.
  • Storage at -20°C, protected from light, for up to one month in solvent, or six months at -80°C.
  • Initial source is plants from the Leguminosae family, specifically Erythrina sigmoidea Hua.
  • Classification includes flavonoids and flavones.
  • Can be dissolved in DMSO at various concentrations for in vitro studies.

Catalog No. 50-004-17348 Supplier Medchemexpress LLC Supplier No. HYN86491MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The product is an active compound suitable for various biochemical research applications.

  • Available in solid form, off-white to light yellow in color.
  • Purity is 98.46%.
  • Storage at -20°C, protected from light, for up to one month in solvent, or six months at -80°C.
  • Initial source is plants from the Leguminosae family, specifically Erythrina sigmoidea Hua.
  • Classification includes flavonoids and flavones.
  • Can be dissolved in DMSO at various concentrations for in vitro studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.