Learn More
Cell Signaling Technology SignalFire™ ECL Reagent, Chemiluminescent Substrate, Western Blot, 50 mL
SDP
Catalog No. NC1275804
Encompass
SignalFire™ ECL Reagent is an enhanced chemiluminescent substrate designed for the detection of picogram amounts of protein via western blot analysis. By utilizing horseradish peroxidase (HRP) to convert luminol in the presence of hydrogen peroxide, this reagent produces an excited intermediate dianion that emits light upon returning to its ground state. Compared to standard substrates, this system offers enhanced signal robustness and an extended output duration of up to 3 hours.
  • Provides high sensitivity for detection of picogram levels of protein
  • Features extended signal output for up to 3 hours
  • Compatible with HRP-conjugated secondary antibodies
  • Requires no further water dilution due to 2X concentrate formulation
  • Stable for 36 months when stored at room temperature
Catalog No. NC1275804 Supplier Cell Signaling Technology Supplier No. 6883P3
SignalFire™ ECL Reagent is an enhanced chemiluminescent substrate designed for the detection of picogram amounts of protein via western blot analysis. By utilizing horseradish peroxidase (HRP) to convert luminol in the presence of hydrogen peroxide, this reagent produces an excited intermediate dianion that emits light upon returning to its ground state. Compared to standard substrates, this system offers enhanced signal robustness and an extended output duration of up to 3 hours.
  • Provides high sensitivity for detection of picogram levels of protein
  • Features extended signal output for up to 3 hours
  • Compatible with HRP-conjugated secondary antibodies
  • Requires no further water dilution due to 2X concentrate formulation
  • Stable for 36 months when stored at room temperature