Learn More
Cell Signaling Technology SignalSilence™ Beta-Catenin siRNA I, Transfection, Human and Mouse, 300 μL
SDP
Catalog No. 502732511
Encompass
SignalSilence™ β-Catenin siRNA I allows researchers to specifically inhibit β-catenin expression using RNA interference, a technique where gene expression is selectively silenced via the delivery of double-stranded RNA molecules. This siRNA is rigorously tested and validated to reduce target protein expression as confirmed by western analysis. Supplied in RNAse-free water, this product is formulated for transfection applications.
  • Specifically inhibits human and mouse β-catenin expression
  • Rigorously tested in-house to verify reduction of target protein expression
  • Monitored for base-by-base coupling efficiency during synthesis
  • Purified by affinity-solid phase extraction
  • Analyzed by mass spectrometry to ensure composition and lot-to-lot consistency
Catalog No. 50-273-2511 Supplier Cell Signaling Technology Supplier No. 6225S
SignalSilence™ β-Catenin siRNA I allows researchers to specifically inhibit β-catenin expression using RNA interference, a technique where gene expression is selectively silenced via the delivery of double-stranded RNA molecules. This siRNA is rigorously tested and validated to reduce target protein expression as confirmed by western analysis. Supplied in RNAse-free water, this product is formulated for transfection applications.
  • Specifically inhibits human and mouse β-catenin expression
  • Rigorously tested in-house to verify reduction of target protein expression
  • Monitored for base-by-base coupling efficiency during synthesis
  • Purified by affinity-solid phase extraction
  • Analyzed by mass spectrometry to ensure composition and lot-to-lot consistency