Learn More
Cell Signaling Technology SignalSilence™ p21 Waf1/Cip1 siRNA I, Human, 300 μL
SDP
Catalog No. 502732341
Encompass
SignalSilence™ p21 Waf1/Cip1 siRNA I enables the specific inhibition of p21 Waf1/Cip1 expression through RNA interference. By introducing double-stranded RNA molecules into cells, this product allows for the selective silencing of gene expression. It has been rigorously tested in-house to ensure effective reduction of protein expression in specified cell lines.
  • Specifically inhibits p21 Waf1/Cip1 expression using RNA interference
  • Validated for human species reactivity
  • Supplied in RNAse-free water
  • Suitable for use in transfection applications
  • Supports research in cell cycle and checkpoint control pathways
Catalog No. 50-273-2341 Supplier Cell Signaling Technology Supplier No. 6456S
SignalSilence™ p21 Waf1/Cip1 siRNA I enables the specific inhibition of p21 Waf1/Cip1 expression through RNA interference. By introducing double-stranded RNA molecules into cells, this product allows for the selective silencing of gene expression. It has been rigorously tested in-house to ensure effective reduction of protein expression in specified cell lines.
  • Specifically inhibits p21 Waf1/Cip1 expression using RNA interference
  • Validated for human species reactivity
  • Supplied in RNAse-free water
  • Suitable for use in transfection applications
  • Supports research in cell cycle and checkpoint control pathways