Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Public Seating™ Signature Science Lab Table, Textured Black Legs
SCI_ED

Catalog No. S83686 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Dimensions (D x W x H):
48 x 24 x 27 to 42 in.
Top Style:
Chemical Resistant
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (D x W x H) Top Style
S83686 48 x 24 x 27 to 42 in. Chemical Resistant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S83686 Supplier National Public Seating™ Supplier No. SLT42448C

missing translation for 'validMainframeMsg'

NPS Signature Frame Adjustable Height Science Lab Tables have perfect chemistry with any science classroom.

  • Round Legs have (2) 3/8 in. Diameter in Textured Black
  • Legs Adjust Smoothly and Easily in 3/4 in. Increments – from 27 to 42 in. Height
  • Adapts for ADA Requirements
  • Easy Assembly, With Pre-Drilled Pilot Holes
  • Patented Mechanism Allows Table to Rise Effortlessly and Automatically Lock At Your Height Setting Requirement
  • Weight Capacity: 500 lb.
  • Oversized Matching Floor Glides are Standard
  • 10 Year Warranty

Specifications

Product Type Lab Table
For Use With (Application) Science Classrooms
Description Resistant to 13 chemicals
Certifications/Compliance MAS2, ANSI BIFMA
Color Black
Dimensions (D x W x H) 48 x 24 x 27 to 42 in.
Load Bearing Capacity (English) 500 lb.
Top Style Chemical Resistant
Warranty 10 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.