Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Signature Series™ Bluing Reagent

Catalog No. 7301L Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
1.32 gal.
4 x 1 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
73-01L 1.32 gal.
22-050-114 1 gal.
22-050-115 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 73-01L Supplier Epredia™ Supplier No. 7301L
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ensure crisp nuclear detail with the ready-to-use Epredia™ Signature Series Bluing Reagent.

  • Buffered to ensure proper alkalinity (pH = 8.0)
  • Specially formulated to prevent tissue separation from the slide
  • Requires only a one-minute water rinse versus conventional bluing reagents which require up-to five minutes

Available in 5 liter packaging

  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging

Specifications

Product Type Bluing Reagent
Quantity 1.32 gal.
pH 8
Form Liquid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.