Learn More
Epredia™ Signature Series™ Clarifier™ 1, 2
Catalog No. 22050117 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 gal.
1.32 gal.
4 x 1 gal.
Product Type:
Clarifier 1
Clarifier 2
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
22-050-117 1 gal. Clarifier 2
74-02L 1.32 gal. Clarifier 2
22-050-118 4 x 1 gal. Clarifier 1
22-050-119 4 x 1 gal. Clarifier 2
22-050-116 1 gal. Clarifier 1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-050-117 Supplier Epredia™ Supplier No. 7402
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Enhance the nuclear definition and clarity of Signature Series hematoxylins with Epredia™ Signature Series Clarifier™ 1, 2.
  • Improves contrast between Nuclei and Cytoplasm
  • Increases cell transparency
  • Removes background staining caused by excessive tissue adhesive
  • Removes unwanted hematoxylin staining from adhesive without affecting nuclear staining
Note:
  • Clarifier 1 is recommended with Hematoxylin 7211 and Hematoxylin 1
  • Clarifier 2 is recommended with Hematoxylin 7212 and Hematoxylin 2
Specifications
Quantity 1 gal.
Product Type Clarifier 2