Learn More
Epredia™ Signature Series™ Clear-Rite 3™
Catalog No. 6905 Shop All Epredia Products
Change view
Click to view available options
Quantity:
4 x 1 gal.
5 Liter
5 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-05 5 gal.
22-046341 4 x 1 gal.
69-15 5 Liter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 69-05 Supplier Epredia™ Supplier No. 6905
Add to Cart
Add to Cart
This item is not returnable. View return policy
Render complete deparaffinization and clearing during the staining process with Epredia™ Signature Series Clear-Rite 3.
  • Very low odor; Benzene-free
  • Use in all tissue processors and automatic stainers
  • Does not produce hard or brittle specimens, even after prolonged exposure
Specifications
Quantity 5 gal.
WARNING: Cancer - www.P65Warnings.ca.gov