Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Signature Series™ Hematoxylin 1

Catalog No. 7221L Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 pt.
1.32 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-21L 1.32 gal.
22-050-112 1 pt.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 72-21L Supplier Epredia™ Supplier No. 7221L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Yield visually similar results to Gills 1 and 2 with this uniquely formulated product.

  • No filtration required
  • Stains up to 2500 slides per pint
  • Part of the Signature Series Staining System
  • Crisp, well-defined nuclear detail chromatin delineation

Available in 5-liter packaging

  • Reduces plastic by 90%
  • Increases stain stability
  • Reduces messy stain spills
  • Saves 35% storage space versus traditional packaging

Specifications

Quantity 1.32 gal.
Product Type Hematoxylin 1
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.