Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ NFPA Hazardous Chemical Label

Catalog No. 19110341 Shop All Brady Corporation Products
Change view
Click to view available options
Adhesive Type:
Permanent Acrylic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Adhesive Type
19-110-341 Permanent Acrylic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-110-341 Supplier Brady™ Supplier No. 121456
  • $33.00
    $30.65 / Pack of 5

    Save $2.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Inform employees of chemical hazards, as required by OSHA's Federal Hazard Communication Standard 29 CFR 1910.1200.

  • Thermal transfer printable flexible vinyl film coated with a permanent pressure sensitive acrylic adhesive
  • -4°F to 167°F
  • Up to 3 years average outdoor durability (Average expected outdoor life of product will depend on user definition of failure, climatic conditions, mounting techniques and material color)
  • Designed to be used with the MINIMARK, Global mark, Power mark and BBP 31 printers (per regional availability) as well as for pre-printed labeling and floor marking
  • Brady B-7569 is used for Pipe Markers, Arrow Tape, Warehouse Marking, Floor Marking, Warning Panels, Safety Signs and Hazardous Substance Identification
  • White, red, yellow, green, blue, orange, black
  • Permanent acrylic adhesive

Specifications

For Use With (Equipment) MINIMARK, Globalmark, Powermark and BBP 31 printers (per regional availability)
Material Vinyl
Adhesive Type Permanent Acrylic
Description Vinyl film with permanent adhesive
Mounting Type Self-adhesive
For Use With (Application) for Pipe Markers, Arrow Tape, Warehouse Marking, Floor Marking, Warning Panels, Safety Signs and Hazardous Substance Identification
Height (English) 7 in.
Height (Metric) 17.7 cm
Max. Temperature (English) 167°F
Min. Temperature (English) -4°F
Thickness 0.004 in.
Width (English) 10 in.
Width (Metric) 25.4 cm
Temperature Range (Metric) -20°C to 75°C
Shape Rectangular
Temperature Range (English) -4°F to 167°F
Languages English
Displayed Text NFPA Hazardous Chemical
Product Type NFPA Placard
Color Red
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.