Learn More
Description
- Visible when cover is removed from confined space entry
- Label affixed to cover is made of indoor/outdoor grade over laminated polyester
- Black/Red on White
- Self Sticking Polyester
- DANGER: CONFINED SPACE PERMIT REQUIRED UNAUTHORIZED ENTRY PROHIBITED... ETC
Specifications
Specifications
| For Use With (Equipment) | Manhole Cover |
| Material | Polyester |
| Adhesive Type | Cold-temperature Pressure-sensitive Permanent Acrylic |
| Description | Self-adhesive Polyester |
| Mounting Type | Self-adhesive |
| Shape | Octagonal |
| Languages | English |
| Displayed Text | DANGER: CONFINED SPACE PERMIT REQUIRED UNAUTHORIZED ENTRY PROHIBITED... ETC |
| Product Type | Confined Space Sign |
| Color | Red on White |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
