Learn More
Medchemexpress LLC Siguazodan | 115344-47-3 | 99.1% | 10 MG
SDP
Catalog No. 5000299190
Encompass_Preferred
Siguazodan is a selective phosphodiesterase 3 (PDE3) inhibitor supplied as a high-purity research reagent. It increases intracellular cAMP, exhibits anti-platelet and vasodilatory activity in preclinical studies, and is provided as a solid suitable for laboratory use.
  • Potent and selective PDE3 inhibitor.
  • High purity (99.1%).
  • Provided as a 10 MG solid for laboratory use.
  • Demonstrated anti-platelet and vasodilatory effects in preclinical studies.
  • Intended for research and analytical applications only.
Catalog No. 50-002-99190 Supplier Medchemexpress LLC Supplier No. HY1902610MG
Siguazodan is a selective phosphodiesterase 3 (PDE3) inhibitor supplied as a high-purity research reagent. It increases intracellular cAMP, exhibits anti-platelet and vasodilatory activity in preclinical studies, and is provided as a solid suitable for laboratory use.
  • Potent and selective PDE3 inhibitor.
  • High purity (99.1%).
  • Provided as a 10 MG solid for laboratory use.
  • Demonstrated anti-platelet and vasodilatory effects in preclinical studies.
  • Intended for research and analytical applications only.