Learn More
Enzo Life Sciences Siguazodan (5mg). CAS: 115344-47-3
SDP
Catalog No. 502011390
Encompass_Preferred
A selective phosphodiesterase PDE III inhibitor, IC50=0.7µM. Siguazodan is a potent inotropic, vasodilator and antiplatelet agent with sustained duration of action. It synergizes with PDE IV inhibitors (roliprams) in blocking antigen-induced bronchospasm in the guinea pig. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: Ambient.
Catalog No. 50-201-1390 Supplier Enzo Life Sciences Supplier No. BMLPD1790005
A selective phosphodiesterase PDE III inhibitor, IC50=0.7µM. Siguazodan is a potent inotropic, vasodilator and antiplatelet agent with sustained duration of action. It synergizes with PDE IV inhibitors (roliprams) in blocking antigen-induced bronchospasm in the guinea pig. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: Ambient.