Learn More
Aviva Systems Biology Corp Sik1 Antibody - middle region (ARP45917_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45917_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Sik1. It was validated on Western Blot. . Alternate names: Sik, Snf1lkApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-285-231