Learn More
Aviva Systems Biology Corp SIK2 Antibody - N-terminal region (ARP62714_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62714_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SIK2. It was validated on Western Blot. . Alternate names: DKFZp434K1115, KIAA0781, LOH11CR1I, QIK, SNF1LK2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-751