Learn More
Cayman Chemical SIKVAV (acetate), Purity: ≥98%, 100 mg
SDP
Catalog No. 502910959
Encompass_Preferred
A laminin α1-derived peptide; binds to α3, α6, and β1 integrins; induces adhesion of CAC2 adenoid cystic carcinoma cells at 1 µg/ml; stimulates the proliferation, migration, and secretion of EGF, VEGF, TGF-β, and FGF in primary mouse fibroblasts at 100 µg/ml; topical administration of a chitosan hydrogel containing SIKVAV promotes wound healing and angiogenesis in mice
Catalog No. 50-291-0959 Supplier Cayman Chemical Supplier No. 35159100 MG
A laminin α1-derived peptide; binds to α3, α6, and β1 integrins; induces adhesion of CAC2 adenoid cystic carcinoma cells at 1 µg/ml; stimulates the proliferation, migration, and secretion of EGF, VEGF, TGF-β, and FGF in primary mouse fibroblasts at 100 µg/ml; topical administration of a chitosan hydrogel containing SIKVAV promotes wound healing and angiogenesis in mice