Learn More
Genscript Corporation Antibody Protein Proteins Proteins 10 UG 1/EA
Supplier:  Genscript Corporation Z0321310
Encompass
Interleukin-6 Receptor (IL-6R) is a single trans-membrane protein that is the receptor for Interleukin-6 (IL-6). IL-6R forms a hexameric complex upon binding 2 molecules of IL-6 and two molecules of glycoprotein 130 (gp130) which activates intracellular JAK/STAT pathways. Although the normal form of IL-6R is the membrane-bound 80 kDa subunit a soluble form of IL-6R (sIL-6R) can be generated physiologically by limited proteolysis or alternative splicing. sIL-6R binds to both IL-6 and gp130 generating intracellular signaling. In the immune system sIL-6R is produced by both naïve and memory CD4 T-cells and strongly augments IL-6 ligands induction of Th-17 cells.
Catalog No. 50-252-9712